Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Chlorododecane for synthesis

Supplier: MilliporeSigma

Synonyms: DodecylchlorideLauryl chloride

8.05340.1000 8.05340.0100 8.05340.1000 8.05340.0100
EM8.05340.1000CS 594.94 USD InStock
EM8.05340.1000 EM8.05340.0100 CA8.05340.1000 CA8.05340.0100
1-Chlorododecane for synthesis
1-Chlorododecane


Formula: C₁₂H₂₅Cl
MW: 204,78 g/mol
Boiling Pt: 260 °C (1013 hPa)
Melting Pt: -9 °C
Density: 0.867 g/cm³ (20 °C)
Storage Temperature: ambient
MDL Number: MFCD00013688
CAS Number: 112-52-7
EINECS: 203-981-5
Order Now
Specification Test Results
Solubility  - 0.007 g/l (25 °C)
Melting Point -9 °C
Molar Mass 204.78 g/mol
Boiling Point 124 °C (13 hPa)
Vapor Pressure  - 0.1 hPa (20 °C)
Flash Point 130 °C
Density 0.87 g/cm3 (20 °C)
Ignition Point 190 °C


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty