Hexafluoroacetone sesquihydrate for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: 1,1,1,3,3,3-Hexafluoroacetone sesquihydrate

M-Clarity™ quality level = MQ200

8.04521.0005 8.04521.0005
EM8.04521.0005CS 297944 USD
EM8.04521.0005 CA8.04521.0005
Hexafluoroacetone sesquihydrate for synthesis, Sigma-Aldrich®
Hexafluoroacetone sesquihydrate
Formula: C₃F₆O·1.5H₂O
MW: 193.05 g/mol
MDL Number: MFCD06800792
UN: 2552
ADR: 6.1,II

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point 11 - 20 °C
Molar Mass 166.02 g/mol
Vapor Pressure 202 hPa (30 °C)
Density 1.69 g/cm3 (20 °C)
pH 1 - 2 (H2O, 20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR