2-Bromophenol for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ200

8.04166.0025 8.04166.0025
EM8.04166.0025CS 18767 USD
CA8.04166.0025 EM8.04166.0025
2-Bromophenol for synthesis, Sigma-Aldrich®
2-Bromophenol
Formula: C₆H₅BrO
MW: 173.01 g/mol
MDL Number: MFCD00002146
CAS Number: 95-56-7
UN: 3082
ADR: 9,III

Order Now

Specification Test Results

Solubility (20 °C) practically insoluble
Melting Point 5 °C
Molar Mass 173.01 g/mol
Boiling Point 195 °C
Flash Point 108 °C
Refractive Index 1.5893 (20 °C, 589 nm)
Density 1.63 g/cm3 (20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR