Germanium(IV) chloride for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
Germanium tetrachloride
, Tetrachlorogermane
M-Clarity™ quality level = MQ200
8.04079.0025
8.04079.0025
EM8.04079.0025CS
48008
USD
EM8.04079.0025
CA8.04079.0025
Germanium(IV) chloride for synthesis, Sigma-Aldrich®
Germanium (IV) chloride
Formula:
Cl₄Ge MW: 214.42 g/mol |
MDL Number:
MFCD00011029 CAS Number: 10038-98-9 UN: 3264 ADR: 8,II |
Specification Test Results
Solubility | (20 °C) Hydrolysis,(decomposition) |
Melting Point | -49.5 °C |
Molar Mass | 214.4 g/mol |
Boiling Point | 83.1 °C |
Vapor Pressure | 101 hPa |
Density | 1.88 g/cm3 (20 °C) |
pH |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...