N,N-Dimethylacetamide for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
Acetic acid dimethylamide
M-Clarity™ quality level = MQ200
8.03235.9026
8.03235.1000
8.03235.2500
8.03235.9026
8.03235.2500
EM8.03235.2500CS
548.26
USD
CA8.03235.2500
CA8.03235.1000
CA8.03235.9026
EM8.03235.2500
EM8.03235.9026
N,N-Dimethylacetamide for synthesis, Sigma-Aldrich®
N,N-Dimethylacetamide
Formula:
CH₃CON(CH₃)₂ MW: 87.12 g/mol Boiling Pt: 166.1 °C (1013 hPa) Melting Pt: –20 °C Density: 0.937 g/cm³ (20 °C) Flash Pt: 70 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008686 CAS Number: 127-19-5 EINECS: 204-826-4 ADR: 6.1,III Merck Index: 13,03254 |
Specification Test Results
Solubility | (20 °C) soluble |
Melting Point | -20 °C |
Molar Mass | 87.12 g/mol |
Boiling Point | 165 - 166 °C (1013 hPa) |
Vapor Pressure | 1.76 hPa (20 °C) |
Flash Point | 64 °C |
Refractive Index | 1.423 (20 °C, 589 nm) |
Explosion Limit | 1.7 - 11.5 %(V) |
Density | 0.94 g/cm3 (20 °C) |
pH | 4 (200 g/l, H2O, 20 °C) |
Ignition Point | 400 °C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...