Isopropenyl acetate for synthesis, Sigma-Aldrich®

Danger


M-Clarity™ quality level = MQ200

8.03138.1000 8.03138.0100 8.03138.1000 8.03138.0100
EM8.03138.1000CS 615.83 USD
CA8.03138.0100 EM8.03138.1000 EM8.03138.0100 CA8.03138.1000
Isopropenyl acetate for synthesis, Sigma-Aldrich®
Isopropenyl acetate
Formula: C₅H₈O₂
MW: 100.12 g/mol
Boiling Pt: 94 °C (1013 hPa)
Melting Pt: –93 °C
Density: 0.9188 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008709
CAS Number: 108-22-5
EINECS: 203-562-7
UN: 2403
ADR: 3,II
Merck Index: 13,05223

Order Now

Specification Test Results

Solubility 34 g/l (20 °C)
Melting Point -93 °C
Molar Mass 100.12 g/mol
Boiling Point 97 °C (1013 hPa)
Vapor Pressure 23 hPa (20 °C)
Flash Point 16 °C
Refractive Index 1.4 (20 °C, 589 nm)
Explosion Limit 1.8 %(V)
Density 0.92 g/cm3 (20 °C)
pH 3 (34 g/l, H2O, 20 °C)
Ignition Point 439 °C DIN 51794

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR