Cyclooctanol for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ200

8.02661.0025 8.02661.0025
EM8.02661.0025CS 4288.46 USD
EM8.02661.0025 CA8.02661.0025
Cyclooctanol for synthesis, Sigma-Aldrich®
Cyclooctanol
Formula: C₈H₁₆O
MW: 128.21 g/mol
Boiling Pt: 203 °C (1013 hPa)
Melting Pt: 15 °C
Density: 0.97 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00001744
CAS Number: 696-71-9
EINECS: 211-800-6

Order Now

Specification Test Results

Solubility 5.4 g/l (20 °C)
Melting Point 15 °C
Molar Mass 128.21 g/mol
Boiling Point 209 °C (1013 hPa)
Vapor Pressure 0.1 hPa
Flash Point 86 °C
Refractive Index 1.485 (20 °C, 589 nm)
Density 0.97 g/cm3 (20 °C)
Ignition Point 300 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR