Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Chloro-4-nitroaniline for synthesis

Supplier: MilliporeSigma

Warning

8.02624.0250 8.02624.0250
EM8.02624.0250CS 661.07 USD InStock
EM8.02624.0250 CA8.02624.0250
2-Chloro-4-nitroaniline for synthesis
2-Chloro-4-nitroaniline


Formula: C₆H₅ClN₂O₂
MW: 172.57 g/mol
Boiling Pt: 326 °C (1013 hPa)
Melting Pt: 105 °C
Density: 1 g/cm³ (20 °C)
Storage Temperature: Ambient
CAS Number: 121-87-9
EINECS: 204-502-2
UN: 2237
ADR: 6.1,III
Order Now
Specification Test Results
Solubility 0.23 g/l (20 °C)
Melting Point 104 - 107 °C
Molar Mass 172.57 g/mol
Bulk Density 850 kg/m3
Boiling Point 320 °C (1013 hPa) decomposes
pH 7 (0.23 g/l, H2O, 20 °C)
Ignition Point 522 °C


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty