Disodium benzene-1,3-disulfonate for synthesis, Sigma-Aldrich®

Warning

Synonyms: 1,3-Benzenedisulphonic acid disodium saltBenzene-1,3-disulphonic acid disodium salt

M-Clarity™ quality level = MQ200

8.02193.0100 8.02193.1000 8.02193.0100 8.02193.1000
EM8.02193.1000CS 606.03 USD
CA8.02193.0100 CA8.02193.1000 EM8.02193.1000 EM8.02193.0100
Disodium benzene-1,3-disulfonate for synthesis, Sigma-Aldrich®
Disodium benzene-1,3-disulphonate
Formula: C₆H₄Na₂O₆S₂
MDL Number: MFCD00007492
CAS Number: 831-59-4
EINECS: 212-606-4

Order Now

Specification Test Results

Solubility 663 g/l (20 °C)
Molar Mass 282.19 g/mol
Bulk Density 860 kg/m3
pH 6.9 (500 g/l, H2O, 20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR