Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ethyl benzoate for synthesis

Warning
Synonyms: Benzoic acid ethyl ester

8.01813.1000 8.01813.1000 8.01813.0250 8.01813.0250
EM8.01813.1000CS 415.08 USD InStock
CA8.01813.1000 EM8.01813.1000 EM8.01813.0250 CA8.01813.0250
Ethyl benzoate for synthesis
Ethyl benzoate


Formula: C₉H₁₀O₂
MW: 150.18 g/mol
Boiling Pt: 415 °C (1013 hPa)
Melting Pt: -35 °C
Density: 1.05 g/cm³ (20 °C)
Flash Pt: 91 °C
Storage Temperature: Ambient
MDL Number: MFCD00009109
CAS Number: 93-89-0
EINECS: 202-284-3
Merck Index: 13,03802
Order Now
Specification Test Results
Solubility 0.5 g/l (20 °C)
Melting Point -35 °C
Molar Mass 150.17 g/mol
Boiling Point 213 - 215 °C (1013 hPa)
Vapor Pressure 0.24 hPa (20 °C)
Flash Point 88 °C
Refractive Index 1.5057 (20 °C, 589 nm)
Explosion Limit 1 %(V)
Density 1.05 g/cm3 (20 °C)
Ignition Point 490 °C

 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty