Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Benzyl(methyl)amine for synthesis

Supplier: MilliporeSigma

Warning
Synonyms: N-MethylbenzylamineN-Benzylmethylamine

8.01681.1000 8.01681.0250 8.01681.0250 8.01681.1000
EM8.01681.1000CS 1115.93 USD InStock
EM8.01681.1000 EM8.01681.0250 CA8.01681.0250 CA8.01681.1000
Benzyl(methyl)amine for synthesis
Benzyl(methyl)amine


Formula: C₆H₅CH₂NHCH₃
MW: 121.18 g/mol
Boiling Pt: ~ 184 °C (1013 hPa)
Melting Pt: -55 °C
Density: 0.94 g/cm³ (20 °C)
Flash Pt: 61 °C
Storage Temperature: Ambient
MDL Number: MFCD00008289
CAS Number: 103-67-3
EINECS: 203-133-4
UN: 2735
ADR: 8,II
https://us.vwr.com/stibo/web/std.lang.all/09/57/2370957.jpg
Order Now
Specification Test Results
Solubility 65 g/l (20 °C)
Melting Point -55 °C
Molar Mass 121.18 g/mol
Boiling Point 184 - 190 °C (1013 hPa)
Vapor Pressure 2.1 hPa (20 °C)
Flash Point 61 °C
Refractive Index 1.5235 (20 °C, 589 nm)
Explosion Limit 1.4 - 7.7 %(V)
Density 0.94 g/cm3 (20 °C)
pH 10 (10 g/l, H2O, 20 °C)
Ignition Point 380 °C


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty