Boron trifluoride - methanol complex 20% in methanol for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.01663.0500 8.01663.0100 8.01663.0500 8.01663.0100
EM8.01663.0100CS 635 USD
EM8.01663.0100 EM8.01663.0500 CA8.01663.0100 CA8.01663.0500
Boron trifluoride - methanol complex 20% in methanol for synthesis, Sigma-Aldrich®
Boron trifluoride methanol complex
Formula: CH₄BF₃O
MW: 99.85 g/mol
Density: 0.89 g/cm³ (20 °C)
Flash Pt: 16 °C
MDL Number: MFCD00071635
CAS Number: 373-57-9
EINECS: 206-766-4
UN: 3286
ADR: 3 (6.1, 8),II

Order Now

Specification Test Results

Solubility (20 °C) (decomposition)
Vapor Pressure 4 hPa (20 °C)
Flash Point 16 °C
Explosion Limit 5.5 %(V) Methanol
Density 0.89 g/cm3 (20 °C)
Ignition Point 420 °C
Ignition Point 420 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR