Pimelic acid for synthesis, Sigma-Aldrich®

Warning

Synonyms: 1,5-Pentanedicarboxylic acidHeptanedioic acid1,7-Heptanedioic acid

M-Clarity™ quality level = MQ200

8.00668.0250 8.00668.0050 8.00668.0050 8.00668.0250
EM8.00668.0050CS 13608.08 USD
EM8.00668.0050 CA8.00668.0050 CA8.00668.0250 EM8.00668.0250
Pimelic acid for synthesis, Sigma-Aldrich®
Pimelic acid
Formula: C₇H₁₂O₄
MW: 160.17 g/mol
Boiling Pt: 354 °C (1013 hPa)
Melting Pt: 101…103 °C
Density: 1.329 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004425
CAS Number: 111-16-0
EINECS: 203-840-8
Merck Index: 12,07585

Order Now

Specification Test Results

Solubility 50 g/l (20 °C)
Melting Point 102 - 105 °C
Molar Mass 160.17 g/mol
Boiling Point 212 °C (13 hPa)
Density 1.329 g/cm3 (20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR