Ethyl butyrate for synthesis, Sigma-Aldrich®

Warning

Synonyms: Butyric acid ethyl esterButanoic acid ethyl ester

M-Clarity™ quality level = MQ200

8.00500.0500 8.00500.0500 8.00500.0100 8.00500.0100
EM8.00500.0500CS 237.54 USD
CA8.00500.0100 EM8.00500.0500 EM8.00500.0100 CA8.00500.0500
Ethyl butyrate for synthesis, Sigma-Aldrich®
Ethyl butyrate
Formula: C₆H₁₂O₂
Boiling Pt: 252 °C (1013 hPa)
Melting Pt: –92 °C
Density: 0.88 g/cm³ (20 °C)
Flash Pt: 26 °C
Storage Temperature: Ambient
MDL Number: MFCD00009394
CAS Number: 105-54-4
EINECS: 203-306-4
UN: 1180
ADR: 3,III
Merck Index: 13,03810

Order Now

Specification Test Results

Solubility 6.2 g/l (20 °C)
Melting Point -98 °C
Molar Mass 116.16 g/mol
Boiling Point 120 - 122 °C (1013 hPa)
Vapor Pressure 17 hPa (20 °C)
Flash Point 26 °C
Refractive Index 1.392 (20 °C, 589 nm)
Density 0.88 g/cm3 (20 °C)
Ignition Point 460 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR