1-(2-Aminoethyl)piperazine for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.00361.0250 8.00361.0250
EM8.00361.0250CS 491 USD
CA8.00361.0250 EM8.00361.0250
1-(2-Aminoethyl)piperazine for synthesis, Sigma-Aldrich®
1-(2-Aminoethyl)piperazine
Formula: C₆H₁₅N₃
MDL Number: MFCD00005971
CAS Number: 140-31-8
EINECS: 205-411-0
UN: 2815
ADR: 8 (6.1),III

Order Now

Specification Test Results

Solubility 1000 g/l (20 °C)
Melting Point -19 °C
Molar Mass 129.20 g/mol
Boiling Point 222 °C (1013 hPa)
Vapor Pressure 0.07 hPa (20 °C)
Flash Point 88 °C
Refractive Index 1.4983 (20 °C, 589 nm)
Explosion Limit 2.1 - 10.5 %(V)
Density 0.99 g/cm3 (20 °C)
pH 12 (100 g/l, H2O, 20 °C)
Ignition Point 315 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR