Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to system maintenance, custom pricing and product availability will not be shown from 7:00pm EST Friday through 6:00pm Sunday. Orders placed will be processed after maintenance is complete.

Fumaroyl chloride for synthesis

Supplier: MilliporeSigma

Synonyms: Fumaryl chloride

8.00272.0050 8.00272.0050
EM8.00272.0050CS 6942.12 USD InStock
EM8.00272.0050 CA8.00272.0050
Fumaroyl chloride for synthesis
Fumaroyl chloride


Formula: C₄H₂Cl₂O₂
MW: 152.96 g/mol
Boiling Pt: 161 °C (1013 hPa)
Melting Pt: -2 °C
Density: 1.41 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000733
CAS Number: 627-63-4
EINECS: 211-005-4
UN: 1780
ADR: 8,II
Order Now
Specification Test Results
Solubility (20 °C) (decomposition)
Melting Point -2 °C
Molar Mass 152.96 g/mol
Boiling Point 161 - 164 °C
Vapor Pressure 1.86 hPa (20 °C)
Flash Point 73 °C
Refractive Index 1.4988 (20 °C, 589 nm)
Density 1.41 g/cm3 (20 °C)


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty