trans-Cinnamoyl chloride for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: (E)-Cinnamoyl chloridetrans-3-Phenylacryloyl Chloride

M-Clarity™ quality level = MQ200

8.00236.0100 8.00236.0100
EM8.00236.0100CS 1462 USD
EM8.00236.0100 CA8.00236.0100
trans-Cinnamoyl chloride for synthesis, Sigma-Aldrich®
trans-Cinnamoyl chloride
Formula: C₉H₇ClO
MW: 166.61 g/mol
MDL Number: MFCD00000732
CAS Number: 17082-09-6
EINECS: 203-065-5
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Solubility (20 °C) (decomposition)
Melting Point 32 - 35 °C
Molar Mass 166.6 g/mol
Boiling Point 251 - 258 °C
Flash Point 110 °C
Density 1.16 g/cm3 (20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR