Octanoyl chloride for synthesis

Supplier: MilliporeSigma

8.00199.0250 8.00199.0250
EM8.00199.0250CS 453.76 USD InStock
CA8.00199.0250 EM8.00199.0250
Octanoyl chloride for synthesis
Octanoyl chloride
Formula: C₈H₁₅ClO
MW: 162.66 g/mol
Boiling Pt: 196 °C (1013 hPa)
Melting Pt: -63 °C
Density: 0.95 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000766
CAS Number: 111-64-8
EINECS: 203-891-6
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Solubility (20 °C) (decomposition)
Melting Point -63 °C
Molar Mass 162.65 g/mol
Boiling Point 195 °C (1013 hPa)
Vapor Pressure 0.33 hPa (20 °C)
Flash Point 82 °C
Refractive Index 1.435 (20 °C, 589 nm)
Density 0.95 g/cm3 (20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR