1-Butyl-1-methylpyrrolidiniumbis[oxalato(2-)-O,O']borate, high purity

Synonyms: 1-BUTYL-1-METHYLPYRROLIDINIUM BIS[OXALATO(2-)]BORATE1-Butyl-1-methylpyrrolidinium bisoxalatoborate

4.91211.0025 4.91211.0025
EM4.91211.0025CS 55331.55 USD InStock
CA4.91211.0025 EM4.91211.0025
1-Butyl-1-methylpyrrolidiniumbis[oxalato(2-)-O,O']borate, high purity
1-Butyl-1-methylpyrrolidiniumbis[oxalato(2-)-O,O']borate
Formula: C₁₃H₂₀BNO₈
MW: 329,12 g/mol
MDL Number: MFCD09038861
CAS Number: 625835-91-8

Order Now

Specification Test Results

Melting Point 55 °C
Molar Mass 329.12 g/mol

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR