Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Butyl-1-methylpyrrolidiniumtris(pentafluoroethyl)trifluorophosphate, high purity

Supplier: MilliporeSigma


4.91084.0100 4.91084.0500 4.91084.0500 4.91084.0100 4.91084.0025 4.91084.0025
EM4.91084.0500CS 25537.96 USD InStock
CA4.91084.0100 EM4.91084.0500 CA4.91084.0500 EM4.91084.0100 CA4.91084.0025 EM4.91084.0025
1-Butyl-1-methylpyrrolidiniumtris(pentafluoroethyl)trifluorophosphate, high purity
1-Butyl-1-methylpyrrolidinium tris(pentafluoroethyl)trifluorophosphate


Order Now
Specification Test Results
Melting Point 4 °C
Molar Mass 587.27 g/mol
Density 1.59 g/cm3
pH 5 (H2O, 20 °C)


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty