1-Butyl-1-methylpyrrolidinium bis(trifluoromethylsulfonyl)imide, high purity, Sigma-Aldrich®
Synonyms:
N-Butyl-N-methylpyrrolidinium bis((trifluoromethyl)sulphonyl)imide
, N-Butyl-N-methylpyrrolidinium bis((trifluoromethyl)sulfonyl)imide
M-Clarity™ quality level = MQ200
4.91046.0100
4.91046.0025
4.91046.0100
4.91046.0025
EM4.91046.0100CS
13325.06
USD
EM4.91046.0100
EM4.91046.0025
CA4.91046.0025
CA4.91046.0100
1-Butyl-1-methylpyrrolidinium bis(trifluoromethylsulfonyl)imide, high purity, Sigma-Aldrich®
1-Butyl-1-methylpyrrolidinium bis(trifluoromethylsulphonyl)imide
Formula:
C₁₁H₂₀F₆N₂O₄S₂ MW: 422.41 g/mol |
MDL Number:
MFCD07784447 CAS Number: 223437-11-4 |
Specification Test Results
Melting Point | -6 °C |
Molar Mass | 422.41 g/mol |
Density | 1.40 g/cm3 |
pH | 5 (H2O, 20 °C) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...