1-Ethyl-3-methylimidazoliumbis(trifluoromethylsulfonyl)imide for synthesis, Sigma-Aldrich®

Danger

Synonyms: 1-Ethyl-3-methylimidazolium bis(trifluoromethylsulphonyl)imide1-ethyl-3-methylimidazolium bis((trifluoromethyl)sulphonyl)imide[EMIIm]1-Ethyl-3-methylimidazolium bis(trifluoromethylsulfonyl)imide1-ethyl-3-methylimidazolium bis((trifluoromethyl)sulfonyl)imide

M-Clarity™ quality level = MQ200

4.90189.0100 4.90189.0025 4.90189.0100 4.90189.0025
EM4.90189.0100CS 23232.72 USD
EM4.90189.0100 CA4.90189.0025 CA4.90189.0100 EM4.90189.0025
1-Ethyl-3-methylimidazoliumbis(trifluoromethylsulfonyl)imide for synthesis, Sigma-Aldrich®
1-Ethyl-3-methylimidazolium bis(trifluoromethylsulphonyl)imide
Formula: C₈H₁₁F₆N₃O₄S₂
MW: 391.32 g/mol
MDL Number: MFCD03788927
CAS Number: 174899-82-2
UN: 2810
ADR: 6.1,III

Order Now

Specification Test Results

Melting Point -15 °C
Molar Mass 391.31 g/mol
Density 1.52 g/cm3 (20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR