1-Ethyl-3-methylimidazoliumtrifluoromethanesulfonate for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: 1-Ethyl-3-methylimidazolium trifluoromethanesulfonate1-Ethyl-3-methylimidazolium trifluoromethanesulphonate

M-Clarity™ quality level = MQ200

4.90061.0100 4.90061.0025 4.90061.0025 4.90061.0100
EM4.90061.0025CS 27271 USD
CA4.90061.0100 CA4.90061.0025 EM4.90061.0025 EM4.90061.0100
1-Ethyl-3-methylimidazoliumtrifluoromethanesulfonate for synthesis, Sigma-Aldrich®
1-Ethyl-3-methylimidazolium trifluoromethanesulphonate
Formula: C₇H₁₁F₃N₂O₃S
MW: 260.24 g/mol
MDL Number: MFCD00210008
CAS Number: 145022-44-2

Order Now

Specification Test Results

Melting Point -12 °C
Molar Mass 260.24 g/mol
Density 1.386 g/cm3 (20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR