Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tetramethylammonium tris(pentafluoroethyl)trifluorophosphate, LiChroTest® for synthesis

Supplier: MilliporeSigma


4.90025.0025 4.90025.0025 4.90025.0100 4.90025.0100
EM4.90025.0025CS 25183.33 USD InStock
EM4.90025.0025 CA4.90025.0025 CA4.90025.0100 EM4.90025.0100
Tetramethylammonium tris(pentafluoroethyl)trifluorophosphate, LiChroTest® for synthesis
Tetramethylammonium tris(pentafluoroethyl)trifluorophosphate


Formula: C₁₀H₁₂F₁₈NP
CAS Number: 394692-83-2
EINECS: 000-000-0
Order Now
Specification Test Results
Melting Point 115 °C
Molar Mass 519.15 g/mol
pH 6 (0.01 g/ml, H2O, 25 °C)


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty