Methyl green zinc chloride salt, Certistain® for microscopy (C.I. 42590), Sigma-Aldrich®

Danger

Synonyms: CI 42590Methyl green zinc chloride double saltMethyl GreenEthyl Green

1.15944.0025 1.15944.0025
EM1.15944.0025CS 10001.9 USD
CA1.15944.0025 EM1.15944.0025
Methyl green zinc chloride salt, Certistain® for microscopy (C.I. 42590), Sigma-Aldrich®
Methyl Green zinc chloride salt
Formula: C₂₇H₃₅BrClN₃·xZnCl₂
MW: 653.24
MDL Number: MFCD00151094
CAS Number: 7114-03-6
UN: 3261
ADR: 8,

Order Now

Specification Test Results

Solubility 70 g/l (20 °C)
Melting Point 300 °C
Molar Mass 653.24 g/mol
Bulk Density 300 kg/m3
pH 4.6 (10 g/l, H2O, 20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR