Methylene blue, Certistain® for microscopy (C.I. 52015), Sigma-Aldrich®
|
Warning
|
Synonyms:
Basic Blue 9
, Swiss Blue
, Methylthioninium chloride
, Methylene blue chloride
, 3,7-Bis(dimethylamino)phenothiazin-5-ium chloride
M-Clarity™ quality level = MQ300
1.15943.0025
1.15943.0100
1.15943.0100
1.15943.0025
EM1.15943.0025CS
6149.98
USD
CA1.15943.0025
CA1.15943.0100
EM1.15943.0025
EM1.15943.0100
Methylene blue, Certistain® for microscopy (C.I. 52015), Sigma-Aldrich®
Methylene blue
Formula:
C₁₆H₁₈ClN₃S MW: 319.86 g/mol |
MDL Number:
MFCD00150008 CAS Number: 61-73-4 |
Specification Test Results
Solubility | 50 g/l (20 °C) |
Melting Point | 180 °C (decomposition) |
Molar Mass | 319.86 g/mol (anhydrous) |
Bulk Density | 400 - 600 kg/m3 |
pH | 3 (10 g/l, H2O, 20 °C) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...