Crystal violet, Certistain® for microscopy (C.I. 42555), Sigma-Aldrich®
|
Danger
|
Synonyms:
Basic Violet 3
, Gentiaverm
, Methylrosanilide chloride
, Hexamethyl Violet
, Vianin
, Gentian violet
, Methyl violet 10B
, CI 42555
, Hexamethyl-p-rosaniline chloride
, N-(4-(Bis(4-(dimethylamino)phenyl)methylene)cyclohexa-2,5-dien-1-ylidene)-N-methylmethanaminium chloride
, 4-[4,4'-Bis(dimethylamino) benzhydrylidene]cyclohexa-2,5-dien-1-ylidene]dimethylammonium chloride
, Dimethyl((di(4-(dimethylamino)phenyl)methylen)-2,5-cyclohexadien-1-yliden)ammoniumchlorid
, Viocid
M-Clarity™ quality level = MQ300
1.15940.0025
1.15940.0025
EM1.15940.0025CS
2967.54
USD
EM1.15940.0025
CA1.15940.0025
Crystal violet, Certistain® for microscopy (C.I. 42555), Sigma-Aldrich®
Crystal Violet
Formula:
C₂₅H₃₀ClN₃ MW: 407.99 g/mol Melting Pt: 189…194 °C Density: 1.19 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011750 CAS Number: 548-62-9 EINECS: 208-953-6 UN: 3077 ADR: 9, |
Specification Test Results
Solubility | 10 g/l (20 °C) |
Melting Point | 189 - 194 °C |
Molar Mass | 407.99 g/mol |
Bulk Density | 220 - 400 kg/m3 |
Density | 1.19 g/cm3 (20 °C) |
pH | 2.5 - 3.5 (10 g/l, H2O, 20 °C) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...