Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Basic fuchsin, Certistain® for microscopy (C.I. 42510)

Supplier: MilliporeSigma

Synonyms: Basic-fuchsinPararosanilineFuchsin basicBasic fuchsin hydrochlorideDiamondfuchsinBasic Red 9(4-(4-Aminophenyl)(4-iminocyclohexa-2,5-dienylidene)methyl)-2-methylaniline hydrochlorideBasic fuchsinCI 42510Magenta ORosaniline hydrochloride

1.15937.0025 1.15937.0025 1.15937.0100 1.15937.0100
EM1.15937.0025CS 18025.29 USD InStock
CA1.15937.0025 EM1.15937.0025 EM1.15937.0100 CA1.15937.0100
Basic fuchsin, Certistain® for microscopy (C.I. 42510)
Basic Fuchsin


Formula: C₂₀H₂₀ClN₃
MW: 337,85 g/mol
Melting Pt: 235 °C
Density: 1.22 g/cm³ (20 °C)
Storage Temperature: ambient
MDL Number: MFCD00012569
CAS Number: 632-99-5
EINECS: 211-189-6
Merck Index: 12,05686
Order Now
Specification Test Results
Solubility 4 g/l (25 °C)
Melting Point 235 °C (decomposition)
Molar Mass 337.85 g/mol
Bulk Density 500 kg/m3
pH 5 - 6 (1 g/l, H2O, 25 °C)


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty