Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to system maintenance, custom pricing and product availability will not be shown from 7:00pm EST Friday through 6:00pm Sunday. Orders placed will be processed after maintenance is complete.

Eosin Y (yellowish), Certistain® for microscopy (C.I. 45380)

Supplier: MilliporeSigma

Synonyms: Eosin yellowishTetrabromofluorescein disodium saltEosin Y disodium saltEosin 225Solvent Red 43Disodium 2-(2,4,5,7-tetrabromo-6-oxido-3-oxoxanthen-9-yl)benzoateField's Stain B2,4,5,7-Tetrabromofluorescein disodium salt

1.15935.0100 1.15935.0100
EM1.15935.0100CS 1835.71 USD InStock
EM1.15935.0100 CA1.15935.0100
Eosin Y (yellowish), Certistain® for microscopy (C.I. 45380)
Eosin Y (yellowish)


Formula: C₂₀H₆Br₄Na₂O₅
MW: 691,86 g/mol
Melting Pt: 295 to 296 °C
Storage Temperature: ambient
MDL Number: MFCD00005040
CAS Number: 17372-87-1
EINECS: 241-409-6
Merck Index: 13,03634
Order Now
Specification Test Results
Solubility 300 g/l (20 °C)
Melting Point 295 - 296 °C
Molar Mass 691.86 g/mol
Bulk Density 710 kg/m3
pH 9.2 (10 g/l, H2O, 20 °C)


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty