Toluidine blue O, Certistain® for microscopy (C.I. 52040), Sigma-Aldrich®

Synonyms: TBO3-Amino-7-(dimethylamino)-2-methylphenothiazin-5-ium chloride3-Amino-7-(dimethylamino)-2-methyl-5-phenothiazinium chlorideBlutene chlorideTolonium chlorideBasic Blue 17

1.15930.0025 1.15930.0025
EM1.15930.0025CS 5540.14 USD
CA1.15930.0025 EM1.15930.0025
Toluidine blue O, Certistain® for microscopy (C.I. 52040), Sigma-Aldrich®
Toluidine blue O
Formula: C₁₅H₁₆ClN₃S
MW: 305.83 g/mol
MDL Number: MFCD00011934
CAS Number: 92-31-9

Order Now

Specification Test Results

Solubility 30 g/l (25 °C)
Molar Mass 305.82 g/mol
Bulk Density 410 kg/m3
pH 2.8 (H2O, 25 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR