Orange G, Certistain® for microscopy (C.I. 16230), Sigma-Aldrich®
Synonyms:
Disodium 7-hydroxy-8-phenylazonaphthalene-1,3-disulphonate
, Acid Orange 10
, Wool Orange 2G
, Orange G sodium salt
, CI 16230
M-Clarity™ quality level = MQ300
1.15925.0025
1.15925.0025
EM1.15925.0025CS
1674.51
USD
EM1.15925.0025
CA1.15925.0025
Orange G, Certistain® for microscopy (C.I. 16230), Sigma-Aldrich®
Orange G
Formula:
C₁₆H₁₀N₂Na₂O₇S₂ MW: 452.38 g/mol |
MDL Number:
MFCD00012457 CAS Number: 1936-15-8 |
Specification Test Results
Solubility | 70 g/l (20 °C) |
Molar Mass | 452.37 g/mol |
Bulk Density | 430 kg/m3 |
pH | 9 (10 g/l, H2O, 20 °C) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...