Dihexyl sodium sulfosuccinate USP for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ200

1.13861.0025 1.13861.0025
EM1.13861.0025CS 78986 USD
CA1.13861.0025 EM1.13861.0025
Dihexyl sodium sulfosuccinate USP for synthesis, Sigma-Aldrich®
Dihexyl sodium sulfosuccinate
Formula: C₁₆H₂₉NaO₇S
MDL Number: MFCD00050501
CAS Number: 3006-15-3

Order Now

Specification Test Results

Solubility 364 g/l (25 °C)
Melting Point 230 °C decomposes
Molar Mass 388.46 g/mol
Bulk Density 180 kg/m3
pH 5 - 7 (50 g/l, H2O, 25 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR