Haemalum (Mayer's) solution for microscopy, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Warning
|
1.09249.2500
1.09249.0500
1.09249.0500
1.09249.1000
1.09249.2500
1.09249.1000
EM1.09249.2500CS
1345.37
USD
CA1.09249.2500
EM1.09249.2500
EM1.09249.0500
CA1.09249.1000
EM1.09249.1000
CA1.09249.0500
Haemalum (Mayer's) solution for microscopy, Sigma-Aldrich®
Haemalum (Mayer's)
Specification Test Results
Solubility | (20 °C) soluble |
Density | 1.05 g/cm3 (20 °C) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...