Lead, foil, EMSURE® for analysis, Thickness ∼0.25 mm, Supelco®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ300

1.07365.0500 1.07365.0500
EM1.07365.0500CS 7008 USD
EM1.07365.0500 CA1.07365.0500
Lead, foil, EMSURE® for analysis, Thickness ∼0.25 mm, Supelco®
Lead
Formula: Pb
MW: 207.2 g/mol
MDL Number: MFCD00134050
CAS Number: 7439-92-1
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Solubility (20 °C) practically insoluble
Melting Point 327 °C
Molar Mass 207.2 g/mol
Boiling Point 1740 °C (1013 hPa)
Density 11.3 g/cm3 (20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR