Niobium(V) oxide

1.06868.0100 1.06868.0100
EM1.06868.0100CS 40653.98 USD
CA1.06868.0100 EM1.06868.0100
Niobium(V) oxide
Niobium (V) oxide
Formula: Nb₂O₅
MW: 265.81 g/mol
MDL Number: MFCD00011128
CAS Number: 1313-96-8

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point 1512 °C
Molar Mass 265.81 g/mol
Density 4.5 g/cm3 (20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR