Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Copper(II) oxide, granular, EMSURE® for analysis

Supplier: MilliporeSigma

Synonyms: Copper oxideCupric oxide

1.02768.0500 1.02768.0500
EM1.02768.0500CS 41034.39 USD InStock
EM1.02768.0500 CA1.02768.0500
Copper(II) oxide, granular, EMSURE® for analysis
Copper (II) oxide


Formula: CuO
MW: 79,55 g/mol
Boiling Pt: 2000 °C (1013 hPa)
Melting Pt: 1326 °C
Density: 4.9 g/cm³ (20 °C)
Storage Temperature: ambient
MDL Number: MFCD00010979
CAS Number: 1317-38-0
EINECS: 215-269-1
UN: 3077
ADR: 9,III
Merck Index: 13,02674
Order Now
Specification Test Results
Solubility (20 °C) insoluble
Melting Point 1326 °C (decomposition)
Molar Mass 79.55 g/mol
Bulk Density 500 kg/m3
Density 6.48 g/cm3 (25 °C)
pH 7 (50 g/l, H2O, 20 °C) (slurry)


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty