Cerium(IV) oxide 1-3 mm for elementary analysis, Supelco®

Supplier: MilliporeSigma

Synonyms: Ceric oxideCerium dioxide

1.02263.0100 1.02263.0100
EM1.02263.0100CS 173327 USD
CA1.02263.0100 EM1.02263.0100
Cerium(IV) oxide 1-3 mm for elementary analysis, Supelco®
Cerium (IV) oxide
Formula: CeO₂
MW: 172.11 g/mol
MDL Number: MFCD00010933
CAS Number: 1306-38-3

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point 2000 °C
Molar Mass 172.11 g/mol
Bulk Density 1280 kg/m3
Density 7.3 g/cm3 (20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR