Malachite green oxalate for microscopy, for microbiology (C.I. 42000), Sigma-Aldrich®

Danger

Synonyms: Bis (N-{4-[[4-(dimethylamino)phenyl](phenyl)methylene]cyclohexa-2,5-dien-1-ylidene}-N-methylmethanaminium) trioxalateN-[4-[[4-(Dimethylamino)phenyl]phenylmethylene]-2,5-cyclohexadien-1-ylidene]-N-methyl-methanaminium, ethanedioate, ethanedioate(2:2:1)Basic Green 4N,N,N',N'-Tetramethyl-4,4'-diaminotriphenylcarbenium oxalate

1.01398.0100 1.01398.0025
CA1.01398.0100 CA1.01398.0025
Malachite green oxalate for microscopy, for microbiology (C.I. 42000), Sigma-Aldrich®
Malachite green oxalate
Formula: C₅₂H₅₄N₄O₁₂
MW: 927.02 g/mol
MDL Number: MFCD00011766
CAS Number: 2437-29-8
UN: 2811
ADR: 6.1,III
https://us.vwr.com/stibo/web/std.lang.all/75/29/2297529.jpg

Order Now

Specification Test Results

Solubility 110 g/l (24 °C)
Melting Point 159 °C
Molar Mass 927.01 g/mol
Bulk Density 400 - 500 kg/m3
pH 2.4 (10 g/l, H2O, 24 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR