Would you like to log in again?
Due to system maintenance, custom pricing and product availability will not be shown from 7:00pm EST Friday through 6:00pm Sunday. Orders placed will be processed after maintenance is complete.
Cel-Gro Tissue Culture Rotator, Thermo Scientific
Supplier: Thermo Scientific
1640Q
62403-968EA 2731.25 USD InStock
62403-968
Cel-Gro Tissue Culture Rotator, Thermo Scientific
Shakers and Mixers Rotating/Rocking Shakers
Mix test tube contents for optimum culture results with the gentle rotating motion of the Thermo Scientific™ Cel-Gro Tissue Culture Rotator, featuring a no-maintenance brushless motor
The single drum has an adjustable rotating angle and speed, while the dual drum has a fixed rotating angle and speed.
Ordering information: Drums sold seperately.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty