Saponin, Calbiochem®

Supplier: MilliporeSigma

Warning

558255-100GM 558255-100GM
80058-630EA 96.93 USD InStock
CA80058-630 80058-630
Saponin, Calbiochem®
Saponin

Saponin is a glycoside used to permeabilize cellular membranes


It exhibits hemolytic activity.

Density: 1.015 to 1.02 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00081981
CAS Number: 8047-15-2
EINECS: 232-462-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR