Three-Bladed Propeller Shaft, Ace Glass Incorporated
Supplier: Ace Glass
13590-02 13590-04
89077-788EA 65.87 USD
89077-788 89077-786
Three-Bladed Propeller Shaft, Ace Glass Incorporated
This stainless steel, three-bladed propeller shaft measures 12" (25.4 cm) in length
2" (51 mm) diameter blades can be twisted to the desired pitch to throw up or down. Blades are firmly riveted to the rod; no nuts or set screws to work loose.
Ordering information: Propeller shaft, stainless, 3 blades, 2" propeller diameter.