Would you like to log in again?
Due to system maintenance, custom pricing and product availability will not be shown from 7:00pm EST Friday through 6:00pm Sunday. Orders placed will be processed after maintenance is complete.
Czapek Dox Agar, HiMedia
Supplier: HiMedia
M075-500G M075-2.5KG M075-5KG M075-5KG M075-2.5KG M075-500G
61000-004EA 115.28 USD InStock
61000-004 CA89222-432 CA89222-434 89222-434 89222-432 CA61000-004
Czapek Dox Agar, HiMedia
Media for Microbiology Dehydrated Media for Microbiology
Czapek Dox Agar is a semisynthetic medium for general cultivation of fungi
Media has a pH of 7.10 - 7.50.
Order Now
ORDER
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty