Stainless Steel Mixing Shafts, Ace Glass Incorporated
Supplier: ACE GLASS, INC.
13598-03 13598-07 13598-05 13598-09
89077-790EA 28.22 USD InStock
89077-790 89077-794 89077-792 89077-796
Stainless Steel Mixing Shafts, Ace Glass Incorporated
Stirrers Overhead Stirrers Stirring Shafts
Mixing Shafts are made of 303/304 Stainless Steel
They will accept agitators outfitted with a set screw.