FLUOR DE LYS® Green SIRT5 Fluorometric Drug Discovery Assay Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
FLUOR DE LYS®
BML-AK514-0001
10033-574EA 912.88 USD
10033-574
FLUOR DE LYS® Green SIRT5 Fluorometric Drug Discovery Assay Kit, Enzo Life Sciences
Assays Protein Assays
FLUOR DE LYS® Green SIRT5 fluorometric drug discovery kit is a sensitive fluorometric assay that measures the lysyl desuccinylase activity of the recombinant human SIRT5.
  • Easy to use kit (two-step) for screening SIRT5 inhibitors/activators; includes enough active enzyme for entire plate
  • Suitable for high-throughput screening (Z’-factors >0.73)
  • Optimal, specific SIRT5 substrate means low enzyme concentration, making ‘hit’ validation easy
FLUOR DE LYS®-Succinyl Green substrate has longer excitation/emission wavelengths, avoiding interference often seen with screening compounds at shorter wavelengths.
Delivery information: Kit contains SIRT5 (Sirtuin 5) (human, recombinant), FLUOR DE LYS®-Succinyl Green, Desuccinylase Substrate, FLUOR DE LYS® Developer Concentrate (20X), Nicotinamide (Sirtuin Inhibitor), Suramin sodium (Sirtuin inhibitor), FLUOR DE LYS®-Green Desuccinylated Standard, Sirtuin Assay Buffer, and half volume 96-well black NBS™ microplate.