Matrix metalloproteinase-9 (MMP-9) Fluorometric Drug Discovery Kit, RED, Enzo Life Sciences
Supplier: Enzo Life Sciences
BML-AK306-0001
10033-760EA 586 USD InStock
10033-760
Matrix metalloproteinase-9 (MMP-9) Fluorometric Drug Discovery Kit, RED, Enzo Life Sciences
Proteins and Peptides
Ordering information: Includes 1 vial MMP-9 enzyme, 1 vial Substrate (OmniMMP™ RED), 1 vial 6’-TAMRA calibration standard, 1 vial control inhibitor (NNGH), 1 bottle (20 ml) assay buffer, 1 black 96-well microplate, Instructions.