Would you like to log in again?
Corning® 96-Well TC-Treated Microplates, Sterile, Corning®
Supplier: Corning
3628
29444-274CS 528.54 USD InStock
29444-274
Corning® 96-Well TC-Treated Microplates, Sterile, Corning®
Microplates Cell Culture Plates
These microplates have flat bottoms, with a total volume of 360 µL
Recommended working volumes are 75 to 200 µL. These microplates are treated for optimal cell attachment. The feature non-reversable lids with condensation rings to reduce contamination. Microplates also feature individual alphanumeric codes for well identification. Sterilized by gamma radiation. Certified nonpyrogenic.
Order Now
ORDER
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty