Would you like to log in again?
62 Series Fluorometers, Jenway
628-001 628-501 627-001
95038-454EA 8821.15 USD InStock
95038-454 95038-456 95038-452
62 Series Fluorometers, Jenway
Spectrophotometers and Spectrometers Fluorescence Spectrophotometers
Jenway offers three fluorimeter models to cover a wide range of applications and fluorscence measurements
With press-to-read operation and Total Energy Transfer (TET) technology the output of the high-energy xenon lamp is maximised and its expected life extended so that it should never need replacing within the normal lifetime of the unit. The high quality optics are complemented by the Intelligent Filter Modules (IFM) that are identified by the unit, generating error messages should the wrong filters be fitted or their positions be incorrect.
Each fluorimeter model offers intuitive operation with a user interface based on logical menus that can be navigated from the simple keypad. Up to 20 methods can be created without restriction and saved for future use. They can also be locked against accidental change by password entry. The permanent time and date tag on every stored reading supports Good Laboratory Practice, while calibration reminders and operator identity can also be entered to support conformance and traceability of results.
The 6280 fluorimeter model is ideal for the most sensitive determinations with emission wavelengths up to 650nm. When higher emission wavelengths are required the 6285 fluorimeter with its red-enhanced detector is the unit of choice. For less sensitive applications and a broader wavelength range up to 1100nm the low cost 6270 fluorimeter unit is more suitable
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty