Would you like to log in again?
Bead Ruptor 24 Elite Bead Mill Homogenizer, Omni International
Supplier: OMNI INTERNATIONAL
19-040E
76000-746EA 10641.26 USD InStock
76000-746
Bead Ruptor 24 Elite Bead Mill Homogenizer, Omni International
Homogenizers Benchtop Homogenizers
The Bead Ruptor Elite is the most advanced and easy to use bead mill homogenizer available. It is specifically designed for grinding, lysing, and homogenization of biological samples prior to molecular extraction.
  • Simultaneously homogenize 24x0.5mL, 24x1.5mL, 24x2mL, 12x7mL, 3x15mL, 6x30mL, 3x50mL or 96-well tube strips
  • Most powerful bead mill on the market
  • Touch screen multilinqual interface
  • Convenient front loading design
  • Customizable memory settings
  • Intergrated lid lock
Using sample tubes pre-filled with a variety of lysing beads, the Bead Ruptor 24 vigorously and uniformly shakes the tubes providing an efficient, consistent, high-yield and quality homogenization usually in less than 40 seconds
The Bead Ruptor 24 Elite is designed with a unique carriage motion to ensure that the intra-tube bead movement reduces swirling and results in the highest bead impact forces of any bead mill on the market. The increased power of the Bead Ruptor 24 Elite decreases processing time and reduces sample heating to maximize sample homogenization efficiency.
It is an ideal solution for releasing DNA, RNA, proteins and small molecules from very tough samples while still retaining molecular integrity.
Certifications: CE compliant.
Ordering information: Accessories not included. Includes the Bead Ruptor 24 Elite Drive Unit, User Manual, and 1-year warranty. The Bead Ruptor 24 requires a tube carriage which are sold separately. Bead Ruptor disposable bead tubes sold seperately.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty