    Sort by:
  • Search Within Results
2-(2-Methylphenyl)ethanol
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
Sort by:
Supplier:  TCI America
Description:   CAS Number: 19819-98-8 MDL Number: MFCD00044241 Molecular Formula: C9H12O Molecular Weight: 136.19 Purity/Analysis Method: 98.0% (GC) Form: C...