    Sort by:
  • Search Within Results
Victoria Pure Blue Bo
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  TCI America
Description:   CAS Number: 2390-60-5 MDL Number: MFCD00011877 Molecular Formula: C33H40ClN3 Molecular Weight: 514.15 Form: Crystal Color: Yellow